1-(5-Fluoro-2-(methylthio)phenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

105680-500MG 105680-1G 105680-5G 105680-25G
77323-386EA 113.3 USD
77323-386 77343-350 77359-480 77371-452
1-(5-Fluoro-2-(methylthio)phenyl)ethanol ≥97%
1-(5-Fluoro-2-(methylthio)phenyl)ethanol

Precaución: for Research Use Only

Formula: C₉H₁₁FOS
Storage Temperature: Ambient
MDL Number: MFCD18891419
CAS Number: 1341549-81-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR