1-Chloro-2-fluoro-4-iodo-3-isopropoxybenzene ≥95%

Supplier: AOBChem USA

Warning

105478-500MG 105478-1G 105478-5G 105478-25G
77323-384EA 198.79 USD
77323-384 77347-450 77359-478 77383-422
1-Chloro-2-fluoro-4-iodo-3-isopropoxybenzene ≥95%
1-Chloro-2-fluoro-4-iodo-3-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₉ClFIO
MW: 314.523 g/mol
Storage Temperature: Ambient
CAS Number: 2918956-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR