5-Iodo-2-methoxy-4-methylphenol ≥97%

Supplier: AOBChem USA

Warning

105392-500MG 105392-1G 105392-5G 105392-25G
77323-382EA 119.48 USD
77323-382 77343-652 77359-476 77371-450
5-Iodo-2-methoxy-4-methylphenol ≥97%
5-Iodo-2-methoxy-4-methylphenol

Precaución: for Research Use Only

Formula: C₈H₉IO₂
Storage Temperature: Ambient
MDL Number: MFCD34778442
CAS Number: 749874-23-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR