2,4-Difluoro-6-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

105148-500MG 105148-1G 105148-5G
77336-770EA 393.46 USD
77336-770 77347-448 77371-446
2,4-Difluoro-6-methoxybenzoic acid ≥97%
2,4-Difluoro-6-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD22415159
CAS Number: 1104386-44-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR