2-Bromo-4-fluoro-3-(trimethylsilyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

105022-500MG 105022-1G 105022-5G 105022-25G
77323-376EA 244.11 USD
77323-376 77347-444 77359-468 77383-418
2-Bromo-4-fluoro-3-(trimethylsilyl)benzaldehyde ≥97%
2-Bromo-4-fluoro-3-(trimethylsilyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrFOSi
MW: 275.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34778295
CAS Number: 2918886-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR