3,4-Dibromo-5-fluoropyridine ≥97%

Supplier: AOBChem USA

Warning

104689-500MG 104689-1G 104689-5G 104689-25G
77323-372EA 111.24 USD
77323-372 77343-292 77359-462 77371-438
3,4-Dibromo-5-fluoropyridine ≥97%
3,4-Dibromo-5-fluoropyridine

Precaución: for Research Use Only

Formula: C5H2Br2FN
MW: 254.882 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13193300
CAS Number: 1260843-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR