(5-Bromo-2-fluoro-4-methoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

104636-1G 104636-5G 104636-25G
77341-724EA 101.97 USD
77341-724 77359-456 77371-434
(5-Bromo-2-fluoro-4-methoxyphenyl)(methyl)sulfane ≥97%
(5-Bromo-2-fluoro-4-methoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈BrFOS
Storage Temperature: Ambient
MDL Number: MFCD34778146
CAS Number: 2090849-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR