1,3-Dibromo-2-methyl-5-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

104601-500MG 104601-1G 104601-5G 104601-25G
77323-368EA 242.05 USD
77323-368 77347-436 77359-454 77383-412
1,3-Dibromo-2-methyl-5-(trifluoromethyl)benzene ≥95%
1,3-Dibromo-2-methyl-5-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₅Br₂F₃
MW: 317.929 g/mol
Storage Temperature: Ambient
CAS Number: 231953-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR