2-Iodo-6-isopropylphenol ≥95%

Supplier: AOBChem USA

Warning

104389-500MG 104389-1G 104389-5G 104389-25G
77323-366EA 231.75 USD
77323-366 77347-434 77359-452 77383-410
2-Iodo-6-isopropylphenol ≥95%
2-Iodo-6-isopropylphenol

Precaución: for Research Use Only

Formula: C₉H₁₁IO
Storage Temperature: Ambient
MDL Number: MFCD16997200
CAS Number: 600738-63-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR