(4-Isopropoxy-3-Isopropylphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

104331-500MG 104331-1G 104331-5G 104331-25G
77323-364EA 226.6 USD
77323-364 77347-432 77359-450 77383-408
(4-Isopropoxy-3-Isopropylphenyl)(methyl)sulfane ≥95%
(4-Isopropoxy-3-Isopropylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₃H₂₀OS
MW: 224.362 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34778077
CAS Number: 2918944-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR