4-Isopropoxy-3-Isopropylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

104327-500MG 104327-1G 104327-5G 104327-25G
77323-362EA 270.89 USD
77323-362 77347-430 77359-448 77383-406
4-Isopropoxy-3-Isopropylbenzoic acid ≥97%
4-Isopropoxy-3-Isopropylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₃
Storage Temperature: Ambient
MDL Number: MFCD28357004
CAS Number: 161070-01-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR