4-(4-Ethoxy-2-fluorophenyl)tetrahydro-2H-pyran-4-ol ≥97%

Supplier: AOBChem USA

Warning

104239-500MG 104239-1G 104239-5G 104239-25G
77323-356EA 140.08 USD
77323-356 77344-764 77359-438 77383-400
4-(4-Ethoxy-2-fluorophenyl)tetrahydro-2H-pyran-4-ol ≥97%
4-(4-Ethoxy-2-fluorophenyl)tetrahydro-2H-pyran-4-ol

Precaución: for Research Use Only

Formula: C₁₃H₁₇FO₃
MW: 240.271 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34778000
CAS Number: 2901109-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR