1-(4-Ethoxy-2-fluorophenyl)cyclohexanol ≥95%

Supplier: AOBChem USA

Warning

104238-500MG 104238-1G 104238-5G 104238-25G
77323-354EA 173.04 USD
77323-354 77346-434 77359-436 77383-398
1-(4-Ethoxy-2-fluorophenyl)cyclohexanol ≥95%

Precaución: for Research Use Only

Formula: C₁₄H₁₉FO₂
Storage Temperature: Ambient
MDL Number: MFCD34777999
CAS Number: 2901083-39-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR