1-(4-Ethoxy-3-fluorophenyl)-4,4-dimethylcyclohexanol ≥97%

Supplier: AOBChem USA

Warning

104207-1G 104207-5G 104207-25G
77340-972EA 64.89 USD
77340-972 77359-432 77371-426
1-(4-Ethoxy-3-fluorophenyl)-4,4-dimethylcyclohexanol ≥97%
1-(4-Ethoxy-3-fluorophenyl)-4,4-dimethylcyclohexanol

Precaución: for Research Use Only

Formula: C₁₆H₂₃FO₂
MW: 266.351 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34777970
CAS Number: 2901085-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR