4-(3-Chloro-4-methylphenyl)tetrahydro-2H-pyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

104120-500MG 104120-1G 104120-25G 104120-5G
77323-350EA 287.37 USD
77323-350 77347-426 77383-396 77371-424
4-(3-Chloro-4-methylphenyl)tetrahydro-2H-pyran-4-ol ≥95%
4-(3-Chloro-4-methylphenyl)tetrahydro-2H-pyran-4-ol

Precaución: for Research Use Only

Formula: C₁₂H₁₅ClO₂
MW: 226.699 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31694058
CAS Number: 1692377-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR