1-(2-Fluoro-3-(methylthio)phenyl)cyclohexanol ≥97%

Supplier: AOBChem USA

Warning

104110-500MG 104110-1G 104110-25G 104110-5G
77323-346EA 301.79 USD
77323-346 77347-422 77383-392 77371-420
1-(2-Fluoro-3-(methylthio)phenyl)cyclohexanol ≥97%
1-(2-Fluoro-3-(methylthio)phenyl)cyclohexanol

Precaución: for Research Use Only

Formula: C₁₃H₁₇FOS
MW: 240.337 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34777882
CAS Number: 2901099-35-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR