Methyl 7-bromo-5-chloro-3-(trifluoromethyl)benzo[b]thiophene-2-carboxylate ≥97%

Supplier: AOBChem USA

Warning

104050-500MG 104050-1G 104050-5G 104050-25G
77323-344EA 153.47 USD
77323-344 77345-500 77359-430 77383-390
Methyl 7-bromo-5-chloro-3-(trifluoromethyl)benzo[b]thiophene-2-carboxylate ≥97%
Methyl 7-bromo-5-chloro-3-(trifluoromethyl)benzo[b]thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₅BrClF₃O₂S
Storage Temperature: Ambient
MDL Number: MFCD34777844
CAS Number: 2901096-08-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR