3,4-Dibromo-2-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

103758-500MG 103758-1G 103758-25G 103758-5G
77323-342EA 298.7 USD
77323-342 77347-420 77383-388 77371-418
3,4-Dibromo-2-methylbenzoic acid ≥97%
3,4-Dibromo-2-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆Br₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34777783
CAS Number: 2383994-37-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR