2-Chloro-5-ethoxy-4-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

103662-500MG 103662-1G 103662-5G
77337-934EA 452.17 USD
77337-934 77347-416 77371-414
2-Chloro-5-ethoxy-4-fluorobenzaldehyde ≥97%
2-Chloro-5-ethoxy-4-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₈ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD34777708
CAS Number: 2385402-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR