1,2-Dibromo-3-fluoro-4-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

103287-500MG 103287-1G 103287-5G 103287-25G
77323-336EA 234.84 USD
77323-336 77347-410 77359-426 77383-384
1,2-Dibromo-3-fluoro-4-methylbenzene ≥95%
1,2-Dibromo-3-fluoro-4-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅Br₂F
Storage Temperature: Ambient
MDL Number: MFCD28784977
CAS Number: 1500635-65-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR