Morpholino(pyridin-4-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

102944-1G 102944-5G 102944-25G
77340-858EA 60.77 USD
77340-858 77359-424 77371-408
Morpholino(pyridin-4-yl)methanone ≥97%
Morpholino(pyridin-4-yl)methanone

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂O₂
MW: 192.214 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00462330
CAS Number: 52482-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR