Pyridin-4-yl(pyrrolidin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

102942-1G 102942-5G 102942-25G
77340-882EA 61.8 USD
77340-882 77359-422 77371-406
Pyridin-4-yl(pyrrolidin-1-yl)methanone ≥97%
Pyridin-4-yl(pyrrolidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂O
MW: 176.215 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03365534
CAS Number: 86542-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR