N-Cyclopropylisonicotinamide ≥97%

Supplier: AOBChem USA

Warning

102941-500MG 102941-1G 102941-5G 102941-25G
77323-334EA 85.49 USD
77323-334 77342-260 77359-420 77371-404
N-Cyclopropylisonicotinamide ≥97%
N-Cyclopropylisonicotinamide

Precaución: for Research Use Only

Formula: C₉H₁₀N₂O
Storage Temperature: Ambient
MDL Number: MFCD00462321
CAS Number: 25764-75-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR