4-(2,3-Difluorophenyl)tetrahydro-2H-pyran-4-ol ≥97%

Supplier: AOBChem USA

Warning

102706-500MG 102706-1G 102706-5G 102706-25G
77323-330EA 202.91 USD
77323-330 77347-406 77359-414 77383-382
4-(2,3-Difluorophenyl)tetrahydro-2H-pyran-4-ol ≥97%
4-(2,3-Difluorophenyl)tetrahydro-2H-pyran-4-ol

Precaución: for Research Use Only

Formula: C₁₁H₁₂F₂O₂
MW: 214.209 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19626902
CAS Number: 1339934-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR