5-Bromo-3-ethoxy-2-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

102343-500MG 102343-1G 102343-5G 102343-25G
77323-322EA 231.75 USD
77323-322 77347-398 77359-404 77383-374
5-Bromo-3-ethoxy-2-methoxypyridine ≥97%
5-Bromo-3-ethoxy-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₈H₁₀BrNO₂
MW: 232.075 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19688668
CAS Number: 909854-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR