5-Bromo-3-isopropoxy-2-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

102340-500MG 102340-1G 102340-5G 102340-25G
77323-320EA 272.95 USD
77323-320 77347-396 77359-402 77383-372
5-Bromo-3-isopropoxy-2-methoxypyridine ≥97%
5-Bromo-3-isopropoxy-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₉H₁₂BrNO₂
MW: 246.101 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21605115
CAS Number: 1241752-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR