2-Fluoro-6-(trifluoromethyl)nicotinic acid ≥97%

Supplier: AOBChem USA

Warning

102251-500MG 102251-1G 102251-5G
77339-932EA 682.89 USD
77339-932 77347-394 77371-396
2-Fluoro-6-(trifluoromethyl)nicotinic acid ≥97%
2-Fluoro-6-(trifluoromethyl)nicotinic acid

Precaución: for Research Use Only

Formula: C₇H₃F₄NO₂
Storage Temperature: Ambient
MDL Number: MFCD16611308
CAS Number: 1105988-42-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR