2-[2-Bromo-5-(phenylmethoxy)phenyl]-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

10221-1G 10221-5G 10221-25G
77341-656EA 98.88 USD
77341-656 77359-400 77371-394
2-[2-Bromo-5-(phenylmethoxy)phenyl]-1,3-dioxolane ≥95%
2-[2-Bromo-5-(phenylmethoxy)phenyl]-1,3-dioxolane

Precaución: for Research Use Only

Formula: C₁₆H₁₅BrO₃
MW: 335.193 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31619354
CAS Number: 173427-53-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR