1-(2,5-Difluorophenyl)ethan-1-ol ≥97%

Supplier: AOBChem USA

Warning

102144-500MG 102144-1G 102144-5G 102144-25G
77323-318EA 84.46 USD
77323-318 77342-210 77359-396 77371-390
1-(2,5-Difluorophenyl)ethan-1-ol ≥97%
1-(2,5-Difluorophenyl)ethan-1-ol

Precaución: for Research Use Only

Formula: C₈H₈F₂O
Storage Temperature: Ambient
MDL Number: MFCD08442989
CAS Number: 3874-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR