4-Borono-2,6-dimethylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

101428-500MG 101428-1G 101428-25G 101428-5G
77323-312EA 292.52 USD
77323-312 77347-390 77383-368 77371-384
4-Borono-2,6-dimethylbenzoic acid ≥95%
4-Borono-2,6-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BO₄
MW: 193.992 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23163288
CAS Number: 1383104-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR