6-Fluoro-2-methoxy-3-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

101351-500MG 101351-1G 101351-5G 101351-25G
77323-306EA 107.12 USD
77323-306 77343-144 77359-388 77371-378
6-Fluoro-2-methoxy-3-(trifluoromethyl)benzoic acid ≥97%
6-Fluoro-2-methoxy-3-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₆F₄O₃
MW: 238.136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27975463
CAS Number: 1823268-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR