4-Bromo-3-fluoro-N-methoxy-N,2-dimethylbenzamide ≥97%

Supplier: AOBChem USA

Warning

101346-500MG 101346-1G 101346-5G 101346-25G
77323-304EA 281.19 USD
77323-304 77347-384 77359-386 77383-364
4-Bromo-3-fluoro-N-methoxy-N,2-dimethylbenzamide ≥97%
4-Bromo-3-fluoro-N-methoxy-N,2-dimethylbenzamide

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrFNO₂
MW: 276.102 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34776504
CAS Number: 2388489-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR