4-Bromo-3-fluoro-2-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

101341-500MG 101341-1G 101341-5G
77323-302EA 310.03 USD
77323-302 77347-382 77371-376
4-Bromo-3-fluoro-2-methylbenzamide ≥97%
4-Bromo-3-fluoro-2-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₇BrFNO
Storage Temperature: Ambient
MDL Number: MFCD32875896
CAS Number: 2092621-82-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR