1,2-Dibromo-3-chloro-5-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

101312-500MG 101312-1G 101312-5G 101312-25G
77323-300EA 223.51 USD
77323-300 77347-380 77359-384 77383-362
1,2-Dibromo-3-chloro-5-methylbenzene ≥95%
1,2-Dibromo-3-chloro-5-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅Br₂Cl
Storage Temperature: Ambient
MDL Number: MFCD26792625
CAS Number: 953794-53-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR