5-Bromo-2-fluoro-4-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

100408-5G 100408-500MG 100408-25G 100408-1G
77359-370EA 508.82 USD
77359-370 77323-294 77371-360 77342-974
5-Bromo-2-fluoro-4-methoxybenzoic acid ≥97%
5-Bromo-2-fluoro-4-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆BrFO₃
Storage Temperature: Ambient
MDL Number: MFCD20486270
CAS Number: 949014-42-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR