Idex® PEEK Tubings, 100 ft. Length

Supplier: Antyila Scientific
02006-24 02006-43 02006-45 02006-41 02006-21
100460-848EA 404 USD
100460-848 100458-078 100458-082 100463-226 100460-852
Idex® PEEK Tubings, 100 ft. Length
Tubing Chromatography Tubing
An excellent replacement for stainless steel tubing.

  • Flexible PEEK tubing is biocompatible and chemically inert
  • Easily cut to desired lengths
  • Smooth internal surface decreases turbulence
  • Color-coded for easier identification

Applications: Chromatography
Characteristics: Unlike stainless steel, PEEK (polyetheretherketone) tubing is flexible and easy to cut. It provides excellent chemical resistance to both organic and inorganic solvents. Tubing withstands pressures to 5000 psi (3000 psi for model 02006-30). PEEK tubing has a very smooth internal surface,More Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR