Idex® Ethylene Tetrafluoroethylene (ETFE) Tubings
Supplier: Antyila Scientific
02014-31 02013-05 02014-72 02017-39 02014-36
100452-542EA 148.75 USD
100452-542 100461-760 100452-570 100452-568 100457-936
Idex® Ethylene Tetrafluoroethylene (ETFE) Tubings
Tubing Chromatography Tubing
Withstands the highest pressures of fluoropolymer tubing.
  • Excellent chemical resistance
  • Max continuous operating temperature of 80 °C (176 °F)
ETFE offers low gas permeability, excellent mechanical stability, and high pressure-holding capabilities. Due to its low gas permeability, ETFE is an ideal choice for connecting the system pump to the vacuum degasser which will help prevent regassing of mobile phase solvents in transit.