Ziath Mohawk Tube Pickers
Supplier: AZENTA US, INC
ZTS-ATRP ZTS-MHK-48 ZTS-MHK-96 ZTS-MHK-LOW
77321-942EA 317.47 USD
77321-942 77321-952 77321-954 77321-956
Ziath Mohawk Tube Pickers
Tubing Selectors
The Mohawk semi-automatic tube selector enables you to pick individual or groups of tubes directly from 96 or 48- well standard SBS-format racks with ease.
  • Can lift up to sixteen individual tubes at a time, making it easy to remove them from the rack and place in a subsidiary rack or other tube device
  • Works with frozen and thawed tubes
  • Fast and reliable tube picking
  • Integrated 1D barcode reader identifies individual racks and the system software then uses this ID to look up the pre-defined picklist; this reduces the risks of the wrong tubes being selected during picking
  • Compatible with other LIMS via Excel® file
Can work in combination with a Ziath Mirage, Express or Cube reader to automatically create picklists and then verify the contents of the subsidiary (or Daughter) racks and reupload their new positions to FreezerPro or to your LIMS via an Excel file.
The Mohawk works out of the box and needs no set up or calibration. In Automatic mode, The Mohawk allows for operation with minimal interaction of the control PC, while in Worklist mode, a file that describes the tubes that are to be selected is used to control the pins.
Operating system on host PC: Windows® 10, 11
Picklist import: csv, text, Excel, xml, json
Certificaciones: CE marked.
Información para pedidos: Low profile lid adapter and a release block are available to enable the Mohawk to operate safely with Azenta Acoustic tubes. Includes Mohawk tube picker instrument, power adaptor, power cable, Mohawk software and license, 1D rack code scanner and bracket, user guide; one picker.
Envase: One tube picker system per case.