VWR® Biopsy Bags
Supplier: VWR International
77270-056 77270-050 77270-054 77270-048
77270-056BX 465.86 USD
77270-056 77270-050 77270-054 77270-048
VWR® Biopsy Bags
Bags Sample Bags
VWR® biopsy bags are specifically designed to ensure easy and effortless opening during the embedding process. Available in multiple sizes in either standard or premium grade.
  • Heat-sealed seams promote fluid movement around the tissue
  • Specially designed to be easy to open, without specimen loss
  • Available in multiple sizes for specimen versatility
Crafted from nylon, VWR® biopsy bags have heat-sealed seams to guarantee unlimited fluid movement around the tissue. The effortless, peel-open design allows for rapid specimen removal, while preventing the loss of small biopsy specimens. Available in a variety of sizes in both standard and premium grade, our nylon mesh biopsy bags provide a comprehensive solution for tissue processing.