J.T.Baker® BAKERBOND® SPE 96-Well Plates
Supplier: Avantor
7090-P30 7090-P60 7091-P10 7091-P30 7091-P60 7211-P10 7225-P60 7466-P10 7466-P30 7466-P60 7020-P10 7020-P30 7020-P60 7086-P10 7086-P30 7086-P60 7087-P10 7087-P30 7087-P60 7090-P10 7211-P30 7211-P60 7225-P10 7225-P30
JT7090-P30EA 390.25 USD
JT7090-P30 JT7090-P60 JT7091-P10 JT7091-P30 JT7091-P60 JT7211-P10 JT7225-P60 JT7466-P10 JT7466-P30 JT7466-P60 JT7020-P10 JT7020-P30 JT7020-P60 JT7086-P10 JT7086-P30 JT7086-P60 JT7087-P10 JT7087-P30 JT7087-P60 JT7090-P10 JT7211-P30 JT7211-P60 JT7225-P10 JT7225-P30
J.T.Baker® BAKERBOND® SPE 96-Well Plates
Microplates Solid Phase Extraction Plates (SPE)
Solid Phase Extraction (SPE) is commonly used as a highly selective sample clean-up or sample enrichment method prior to subsequent analyses. J.T.Baker® BAKERBOND® SPE 96 well plates are well suited for high throughput and automated SPE methods. With complementary high capacity sorbents to existing spe extraction columns in the J.T.Baker® BAKERBOND® range, higher throughput methodologies can now be simply developed and automated.
  • 96 well format for multiple sample preparation
  • Consistent sorbent weight to reduce variability
  • Different chemistries available to suit a range of analytes
  • Ideal prior to LC-MS, UHPLC or HPLC applications
  • Vacuum manifold can be used (8183-VM)
  • Multi-channel pipettors are recommended
Automated SPE can dramatically speed up sample preparation or enrichment compared to the use of single cartridges or other techniques such as liquid-liquid extraction.