Inkjet Label and Overlaminate Kits
Supplier: Brady Worldwide
175454 175455 175456 175457 175458 175459 175460 175445 175446 175447 175448 175449 175450 175451 175452 175453
77282-318KT 576.91 USD
77282-318 77282-320 77282-322 77282-324 77282-326 77282-328 77282-330 77282-300 77282-302 77282-304 77282-306 77282-308 77282-310 77282-312 77282-314 77282-316
Inkjet Label and Overlaminate Kits
Labels
Take inkjet printing further and label faster with these perfectly sized label and overlaminate kits.
  • Permanent acrylic adhesive
  • Matte finish
  • Inkjet print-receptive material
  • Temperature-resistant, UV-resistant, and water-resistant
  • Service temperature: −40 to 140 °F
Just peel, adhere, and apply with these pre-sized labels and overlaminate. Inkjet-printable label is outfitted with a specially formulated topcoat that invigorates colors and label appearance. Label topcoat also protects against smudges, smears, and splotches. Acrylic overlaminate provides up to 10 years of outdoor durability through improved UV and abrasion resistance. Overlaminate is oversized by a quarter inch for easier application and complete coverage.
An ink cartridge is required to print label material; use part number J20-CMY for J2000 printer, J40-CMY for the J4000 printer, or J50-CMYK for the J5000 printer.