2-Amino-3-bromo-5-chlorophenol 97%

Supplier: Ambeed
Warning

A848948-1G A848948-100MG
77308-448EA 278 USD
77308-448 77784-225
2-Amino-3-bromo-5-chlorophenol 97%
2-Amino-3-bromo-5-chlorophenol

Precaución: for Research Use Only

Formula: C₆H₅BrClNO
MDL Number: MFCD28975091
CAS Number: 1549689-88-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR