Copper(II) bromide 95%

Supplier: Ambeed
Danger

A669946-25G A669946-100G
77308-086EA 24 USD
77308-086 77602-700
Copper(II) bromide 95%
Copper(II) bromide

Precaución: for Research Use Only

Formula: Br₂Cu
MDL Number: MFCD00010970
CAS Number: 7789-45-9
UN: 3260
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR