(3R,4S)-4-(4-Fluorobenzamido)-6-(4-(oxetan-3-yl)piperazin-1-yl)chroman-3-yl methylcarbamate ≥98%

Supplier: Ambeed
Warning

Synonyms: LY 3000328

A473978-10MG A473978-1MG A473978-25MG A473978-50MG A473978-5MG
77307-502EA 149 USD
77307-502 77307-504 77307-506 77307-508 77307-510
(3R,4S)-4-(4-Fluorobenzamido)-6-(4-(oxetan-3-yl)piperazin-1-yl)chroman-3-yl methylcarbamate ≥98%
(3R,4S)-4-(4-Fluorobenzamido)-6-(4-(oxetan-3-yl)piperazin-1-yl)chroman-3-yl methylcarbamate

Precaución: for Research Use Only

Formula: C₂₅H₂₉FN₄O₅
MDL Number: MFCD28502827
CAS Number: 1373215-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR