Diamond® RealSeal™ PETG Media Bottles
Supplier: Globe Scientific
7610030 7610060 7610125 7610250 7610500 7611000 7612000
77308-586CS 224.32 USD
77308-586 77308-588 77308-590 77308-592 77308-594 77308-596 77308-598
Diamond® RealSeal™ PETG Media Bottles
Bottles Plastic Bottles
Diamond® RealSeal™ PETG Square Media Bottles are an ideal solution for protecting media, laboratory reagents, and buffers during storage and transport.
  • ISO 9001 and ISO 13485 production facilty produced and packaged in ISO Class 8 cleanroom
  • Medical grade resins to meet demanding requirements
  • Leakproof, linerless caps for secure closing
  • Gamma sterilized
  • PETG bottle material protects contents during storage
  • Limits transmission of O₂ and CO₂ to protect pH sensitive solutions
Diamond® RealSeal™ PETG Square Media Bottles protect media, laboratory reagents, and buffers during storage and transport. Perfect for cold storage, the PETG bottles remain shatterproof even at −40 °C. Low transmission rates of O₂ and CO₂ maintain optimum pH values for solutions in storage. Bottles include white HDPE caps with integrated seal rings and matching semi-buttressed threads for leakproof performance to 95kPa. Bottles are provided sterile and ready to fill with tamper-evident shrink bands in tray packs for convenience.