Anti-KLH Mouse Monoclonal Antibody [clone: KT85]

Supplier: ANTIBODIES.COM LLC
A131-200 A131-100
77041-498EA 647.3 USD
77041-498 77041-496
Anti-KLH Mouse Monoclonal Antibody [clone: KT85]
Antibodies
Mouse monoclonal (KT85) antibody to KLH for Isotype Control.

  • Validated Applications: Isotype Control
  • Host: Mouse
  • Clonality: Monoclonal
  • Isotype: IgG3
  • Conjugate: Unconjugat

Type: Primary
Antigen: KLH
Clonality: Monoclonal
Clone: KT85
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR