Anti-BIO Tag Rat Monoclonal Antibody [clone: KT89]

Supplier: ANTIBODIES.COM LLC
A104-200 A104-100
77041-408EA 647.3 USD
77041-408 77041-406
Anti-BIO Tag Rat Monoclonal Antibody [clone: KT89]
Antibodies
Rat monoclonal (KT89) antibody to BIO tag for IP with samples derived from Human.

  • Validated Applications: IP
  • Recommended Dilutions: ELISA: 1 µg/ml, WB: 1/500 to 1/10000, IP: 1/100 to 1/1000

Type: Primary
Antigen: BIO Tag
Clonality: Monoclonal
Clone: KT89
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR