Anti-CD16 Chimera Recombinant Monoclonal Antibody [clone: DMC219]

Supplier: ANTIBODIES.COM LLC
A318798-100 A318798-500
77187-830EA 703.3 USD
77187-830 77187-832
Anti-CD16 Chimera Recombinant Monoclonal Antibody [clone: DMC219]
Antibodies
Recombinant chimeric monoclonal [DMC219] antibody to CD16 for ELISA and flow cytometry with samples derived from human.

  • Validated applications: ELISA, Flow cytometry
  • Recommended dilutions: ELISA: 1:5000 to 10000, Flow cytometry: 1:100

Type: Primary
Antigen: CD16
Clonality: Recombinant
Clone: DMC219
Conjugation: Unconjugated
Epitope:
Host: Recombinant
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR