Anti-Asialoglycoprotein Receptor 1/HL-1 Recombinant Recombinant Antibody [clone: DMC688]

Supplier: ANTIBODIES.COM LLC
A318680-100 A318680-500
77187-358EA 703.3 USD
77187-358 77187-360
Anti-Asialoglycoprotein Receptor 1/HL-1 Recombinant Recombinant Antibody [clone: DMC688]
Antibodies
Recombinant chimeric monoclonal [DMC688] antibody to Asialoglycoprotein Receptor 1/HL-1 for flow cytometry with samples derived from human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: 1:100

Type: Primary
Antigen: Asialoglycoprotein Receptor 1/HL-1
Clonality: Recombinant
Clone: DMC688
Conjugation: Unconjugated
Epitope:
Host: Recombinant
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR