Anti-Gastric Mucin Mouse Monoclonal Antibody [clone: ABT-MUC6]

Supplier: ANTIBODIES.COM LLC
A100017-50 A100017-100
77063-594EA 356.8 USD
77063-594 77063-178
Anti-Gastric Mucin Mouse Monoclonal Antibody [clone: ABT-MUC6]
Antibodies
Mouse monoclonal (ABT-MUC6) antibody to MUC6 for IHC with samples derived from Human.

  • Validated Applications: IHC

Type: Primary
Antigen: Gastric Mucin
Clonality: Monoclonal
Clone: ABT-MUC6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR